Alfa Aesar™ Indium(III) isopropoxide, 99.9% (metals basis)

Precursor used in the preparation of indium oxide containing films

Manufacturer: Alfa Aesar™ 4209809

Quantity 10g

 View more versions of this product

Catalog No. AA4209809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38218-24-5
    Linear Formula In[OCH(CH3)2]3
    Formula Weight 292.08
    MDL Number MFCD00210318
    UN Number UN3181
    Chemical Name or Material Indium(III) isopropoxide

    Specifications

    Formula Weight 292.08
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Powder
    Quantity 10g
    Sensitivity Moisture Sensitive
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Storage Note 1
    • Packaged under argon
    UN Number UN3181
    Health Hazard 1 Warning
    Linear Formula In[OCH(CH3)2]3
    MDL Number MFCD00210318
    CAS 38218-24-5
    Melting Point 145°C (dec.)
    Color White
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Chemical Name or Material Indium(III) isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    • Packaged under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit