Learn More
Indium(III) nitrate hydrate, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C-H2O |
Color | White |
Physical Form | Lump/Powder |
Quantity | 10 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water,methanol and ethanol. |
IUPAC Name | indium(3+);trinitrate;hydrate |
Formula Weight | 300.83 (Anhydrous) |
Sensitivity | Hygroscopic |
Appearance | White to off-white lump/powder |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319 |
Precautionary Statement | P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: III |
EINECSNumber | 237-393-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.