Alfa Aesar™ Indium(III) oxide, nanopowder, 99.5% min (metals basis)

Manufacturer: Alfa Aesar™ 4313706

Quantity 5g

 View more versions of this product

Catalog No. AA4313706

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1312-43-2
    Molecular Formula In2O3
    Formula Weight 277.64
    MDL Number MFCD00011060
    Merck Index 14,4952
    Chemical Name or Material Indium (III) oxide

    Specifications

    Formula Weight 277.64
    Molecular Formula In2O3
    CAS 1312-43-2
    Odor Odorless
    Particle Size <50nm APS
    Merck Index 14,4952
    Density 7.18g/mL
    TSCA TSCA
    Physical Form Nanopowder
    MDL Number MFCD00011060
    Melting Point 850°C (volat.)
    Quantity 5g
    Solubility Information Insoluble in water; Soluble in hot mineral acids
    Chemical Name or Material Indium (III) oxide
    Assay ≥99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit