Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indium(III) perchlorate octahydrate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1164218

Quantity 50g

 View more versions of this product

Catalog No. AA1164218


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13465-15-1
MFCD00150241
Indium(III) perchlorate octahydrate
557.30 (413.18 Anhydrous)
UN1481

Specifications

13465-15-1
557.30 (413.18 Anhydrous)
UN1481
Hygroscopic
50g
99.9%, (metals basis)
Crystalline
MFCD00150241
Odorless
800°C
Indium(III) perchlorate octahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit