Alfa Aesar™ Indium(III) phosphide, 99.9999% (metals basis)

Manufacturer: Alfa Aesar™ 1397403

Quantity 1g

 View more versions of this product

Catalog No. AA1397403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22398-80-7
    Formula Weight 145.79
    MDL Number MFCD00016153
    Merck Index 14,4953
    Chemical Name or Material Indium(III) phosphide

    Specifications

    Formula Weight 145.79
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number NL1800000
    Physical Form Pieces
    MDL Number MFCD00016153
    CAS 22398-80-7
    Merck Index 14,4953
    Melting Point 1070°C
    Quantity 1g
    Health Hazard 1 Warning
    Density 4.81g/mL
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    Chemical Name or Material Indium(III) phosphide
    Assay 99.9999%, (metals basis)

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit