Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Indium(III) sulfate, anhydrous, 99.99% (metals basis)
Click to view available options
Quantity:
25g
100g
Specifications
Solubility Information | Soluble in water. |
Formula Weight | 517.83 |
Physical Form | Solid |
Percent Purity | 99.99% |
Density | 3.438 g/mL |
Odor | Odorless |
Sensitivity | Hygroscopic |
Color | Gray |
Quantity | 100g |
Chemical Name or Material | Indium(III) sulfate, Anhydrous |
Show More |
Safety and Handling
EINECSNumber : 236-689-1
RTECSNumber : NL1925000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen