Alfa Aesar™ Indium(III) sulfate hydrate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4287009

Quantity 10g

 View more versions of this product

Catalog No. AA4287009

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula In2(SO4)3·xH2O
    Formula Weight 517.83 (Anhydrous)
    MDL Number MFCD00149747
    Chemical Name or Material Indium (III) sulfate hydrate

    Specifications

    Formula Weight 517.83 (Anhydrous)
    Melting Point 250°C (dec.)
    TSCA TSCA
    Molecular Formula In2(SO4)3·xH2O
    Quantity 10g
    Physical Form Crystalline
    MDL Number MFCD00149747
    Chemical Name or Material Indium (III) sulfate hydrate
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit