Alfa Aesar™ Indium(III) sulfide, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 4483609

Quantity 10g

 View more versions of this product

Catalog No. AA4483609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12030-24-9
    Molecular Formula In2S3
    Formula Weight 325.83
    MDL Number MFCD00011061
    Chemical Name or Material Indium (III) sulfide

    Specifications

    Formula Weight 325.83
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture Sensitive
    Quantity 10g
    Molecular Formula In2S3
    MDL Number MFCD00011061
    Color Red-orange
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Odor Like rotten eggs
    Physical Form Powder
    CAS 12030-24-9
    Melting Point 1050°C
    Chemical Name or Material Indium (III) sulfide
    Assay 99.995%, (metals basis)

    GHS H Statement
    H302-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit