Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) trifluoromethanesulfonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
128008-30-0
C3F9InO9S3
562.03
MFCD00144478
UCYRAEIHXSVXPV-UHFFFAOYSA-K
indium iii trifluoromethanesulfonate,indium triflate,indium trifluoromethanesulphonate,indium 3+ tritriflate,indium 3+ ; trifluoromethanesulfonate,indium iii triflate,indium iii trifluoromethanesulphonate,acmc-209bda,in tfa 3,ksc491q8t
2734680
indium(3+);trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]
128008-30-0
C3F9InO9S3
562.03
MFCD00144478
UCYRAEIHXSVXPV-UHFFFAOYSA-K
indium iii trifluoromethanesulfonate,indium triflate,indium trifluoromethanesulphonate,indium 3+ tritriflate,indium 3+ ; trifluoromethanesulfonate,indium iii triflate,indium iii trifluoromethanesulphonate,acmc-209bda,in tfa 3,ksc491q8t
2734680
indium(3+);trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]
Infrared Spectrum Authentic
Quantity 25g
Color White
Elemental Analysis 97.5% min. (ex Fluor)
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 562.03
Physical Form Powder and Granules
Percent Purity 98%
Chemical Name or Material Indium(III) trifluoromethanesulfonate

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger