Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9995% (metals basis)

Click to view available options
Quantity:
50g
5 x 50g
7440-74-6
In
114.818
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
7440-74-6
In
114.818
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
Quantity 50g
Density 7.310 g/mL
Odor Odorless
Formula Weight 114.818
Physical Form Polycrystalline Ingot
Percent Purity 99.9995%
Grade Puratronic™
Melting Point 156.6°C
Boiling Point 2072°C
Chemical Name or Material Indium ingot
Assay (metals basis)
Show More Show Less

EINECSNumber : 231-180-0

TSCA : Yes

Recommended Storage : Ambient temperatures