Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indium Wire, 0.25mm (0.01 in.) dia., Puratronic™, >99.998% (Metals basis), Alfa Aesar™
Click to view available options
Quantity:
10m
2.5m
25m
Specifications
Quantity | 25m |
Appearance | Silvery-white wire |
Assay Percent Range | >99.998% (Metals basis) |
Odor | Odorless |
Physical Form | Wire, ≈0.36g/m |
Color | Silver-White |
Chemical Name or Material | Indium |
Safety and Handling
EINECSNumber : 231-180-0
TSCA : Yes
Recommended Storage : Ambient temperatures