Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998+% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 11462G5

Quantity 5m

 View more versions of this product

Catalog No. AA11462G5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-74-6
    MDL Number MFCD00134048
    Chemical Name or Material Indium wire, 1.0mm (0.04 in.) dia, Puratronic™

    Specifications

    Quantity 5m
    TSCA TSCA
    Density ≈5.74g/m
    MDL Number MFCD00134048
    CAS 7440-74-6
    Odor Odorless
    Chemical Name or Material Indium wire, 1.0mm (0.04 in.) dia, Puratronic™
    Assay ≥99.998%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit