Learn More
Troemner™ Individual Analytical Precision Ultraclass Gold Weights, with NVLAP Certificate
The weight tolerance of UltraClass Gold weights is equal to ANSI/ASTM E617 Class 00 and OIML R 111 Class E1. They are used as a reference standard for calibrating other reference weights and for calibrating high-precision analytical balances with a readability as low as 0.001mg.
$513.50 - $4850.00
Specifications
Classification UltraClass Gold
Certifications/Compliance NVLAP Accredited Certificate
View More Specs
Products 14
Catalog Number Mfr. No. Weight (Metric) Shape Material Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Shape Material Price Quantity & Availability  
02-214-551 View Documents Troemner™
7009UCGW
10kg Two-piece, Cylindrical with handling knob 316 Stainless Steel Each for $4,850.00
Add to cart
02-214-555 View Documents Troemner™
7017UCGW
100g Two-piece, Cylindrical with handling knob Troemner Alloy 8 Each for $587.00
Add to cart
02-214-556 View Documents Troemner™
7018UCGW
50g Two-piece, Cylindrical with handling knob Troemner Alloy 8 Each for $513.50
Add to cart
02-214-557 View Documents Troemner™
7021UCGW
10g Two-piece, Cylindrical with handling knob Troemner Alloy 8 Each for $513.50
Add to cart
02-214-558 View Documents Troemner™
7022UCGW
5g Two-piece, Cylindrical with handling knob Troemner Alloy 8 Each for $513.50
Add to cart
02-214-559 View Documents Troemner™
7024UCGW
2g Two-piece, Cylindrical with handling knob Troemner Alloy 8 Each for $513.50
Add to cart
02-214-560 View Documents Troemner™
7026UCGW
500mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-561 View Documents Troemner™
7028UCGW
200mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-563 View Documents Troemner™
7030UCGW
50mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-564 View Documents Troemner™
7032UCGW
20mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-565 View Documents Troemner™
7033UCGW
10mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-566 View Documents Troemner™
7034UCGW
5mg One-piece, Flat with one end turned up 304 Stainless Steel Each for $809.50
Add to cart
02-214-567 View Documents Troemner™
7036UCGW
2mg One-piece, Flat with one end turned up 3003-H14 Aluminum Each for $809.50
Add to cart
02-214-568 View Documents Troemner™
7037UCGW
1mg One-piece, Flat with one end turned up 3003-H14 Aluminum Each for $809.50
Add to cart
Description
Description
  • Used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Supplied in high quality polycarbonate case with red insert
  • Manufactured in the United States
The Troemner NVLAP Accredited Certificate provides compliance in both measurement process and data reported to the customer to meet a variety of standards including NVLAP Handbook 150-2, ISO/IEC 9000, ISO/IEC 17025, FDA, GMP. GLP, DOD, ANSI/NCSL Z540-1 and nuclear requirements.
Troemner's NVLAP Laboratory Code is 105013-0. The NIST administered National Voluntary Laboratory Accreditation Program (NVLAP) approves, through periodic audits, all processing and weighing procedures, as well as certificate format and content.
Calibration procedures vary by tolerance class requested. For example, for higher accuracy classes such as Troemner UltraClass Series, ANSI/ASTM E617 Class 000, 00, 0 and 1, OIML R 111 Class E0, E1, E2 and F1, Troemner performs a multiple weighing procedure utilizing standards to determine the mass of a weight.
Made in the USA.
Certifications
NVLAP accreditation
Specifications
Specifications
UltraClass Gold
NVLAP Accredited Certificate