Learn More
Individual aroclor Spex CertiPrep
Click to view available options
Quantity:
1 mL
Aroclor 1268 Individual Semivolatile in Hexane, 1 mL, 200 µg/mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
  • Individual Aroclor 1268
  • Semivolatile Standards
  • 1 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Quantity 1 mL