Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoine Blue, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
4569-88-4
C30H24ClN5O
506.006
SCMDRBZEIUMBBQ-UHFFFAOYSA-N
9575841
(1Z)-1-[(8-amino-3,7-dimethyl-10-phenylphenazin-10-ium-2-yl)hydrazinylidene]naphthalen-2-one;chloride
CC1=CC2=C(C=C1N)[N+](=C3C=C(C(=CC3=N2)C)NN=C4C(=O)C=CC5=CC=CC=C54)C6=CC=CC=C6.[Cl-]
4569-88-4
C30H24ClN5O
506.006
SCMDRBZEIUMBBQ-UHFFFAOYSA-N
9575841
(1Z)-1-[(8-amino-3,7-dimethyl-10-phenylphenazin-10-ium-2-yl)hydrazinylidene]naphthalen-2-one;chloride
CC1=CC2=C(C=C1N)[N+](=C3C=C(C(=CC3=N2)C)NN=C4C(=O)C=CC5=CC=CC=C54)C6=CC=CC=C6.[Cl-]
Infrared Spectrum Authentic
Lambda Max 595 to 601nm (0.001% in methanol)
Loss on Drying 10% max. (110°C, 1 h)
Formula Weight 506.01
Packaging Glass bottle
Quantity 25g
Chemical Name or Material Indoine Blue