Alfa Aesar™ Indole-2-carboxaldehyde, 97%

Manufacturer: Alfa Aesar™ H5463803

Quantity 1g

 View more versions of this product

Catalog No. AAH5463803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19005-93-7
    Molecular Formula C9H7NO
    Formula Weight 145.16
    MDL Number MFCD03001425
    Chemical Name or Material Indole-2-carboxaldehyde

    Specifications

    Formula Weight 145.16
    Molecular Formula C9H7NO
    MDL Number MFCD03001425
    CAS 19005-93-7
    Health Hazard 1 Warning
    Melting Point 138° to 142°C
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material Indole-2-carboxaldehyde
    Assay 97%

    Description

    Applications
    Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

    Solubility
    Soluble in Methanol.

    Notes
    Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit