Alfa Aesar™ Indole-3-acetic acid, 98+%

Manufacturer: Alfa Aesar™ A1055606

Quantity 5g

 View more versions of this product

Catalog No. AAA1055606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87-51-4
    Molecular Formula C10H9NO2
    Formula Weight 175.19
    MDL Number MFCD00005636
    Beilstein 143358
    Merck Index 14,4964
    Synonym Heterauxin; Indolyl-3-acetic acid
    Chemical Name or Material Indole-3-acetic acid

    Specifications

    Formula Weight 175.19
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C10H9NO2
    MDL Number MFCD00005636
    Beilstein 143358
    Merck Index 14,4964
    Synonym Heterauxin; Indolyl-3-acetic acid
    Health Hazard 1 Warning
    Quantity 5g
    Recommended Storage 2°C to 8°C
    Melting Point ≈167°C (dec.)
    Flash Point 171°C (339°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number NL3150000
    CAS 87-51-4
    Chemical Name or Material Indole-3-acetic acid
    Assay ≥98%

    Description

    Applications
    It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

    Solubility
    Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.

    Notes
    Light sensitive. Store at -20°C. ncompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit