Alfa Aesar™ Indole-4-boronic acid pinacol ester, 97+%

Manufacturer: Alfa Aesar™ H52325MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52325MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 388116-27-6
    Molecular Formula C14H18BNO2
    Formula Weight 243.11
    MDL Number MFCD08689896
    Chemical Name or Material Indole-4-boronic acid pinacol ester

    Specifications

    Formula Weight 243.11
    Molecular Formula C14H18BNO2
    MDL Number MFCD08689896
    CAS 388116-27-6
    Quantity 250mg
    Chemical Name or Material Indole-4-boronic acid pinacol ester
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit