Alfa Aesar™ Indole-4-carboxaldehyde, 97%

Manufacturer: Alfa Aesar™ H28019MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28019MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1074-86-8
    Molecular Formula C9H7NO
    Formula Weight 145.16
    MDL Number MFCD01632221
    Synonym 4-Formylindole
    Chemical Name or Material Indole-4-carboxaldehyde

    Specifications

    Formula Weight 145.16
    Molecular Formula C9H7NO
    Synonym 4-Formylindole
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD01632221
    CAS 1074-86-8
    Health Hazard 1 Warning
    Melting Point 143° to 144°C
    Chemical Name or Material Indole-4-carboxaldehyde
    Assay 97%

    Description

    Applications
    4-Indolecarbaldehyde is a synthetic intermediate used for pharmaceutical synthesis. Also used as reactant in Biginelli reaction, synthesis of aurora kinase A inhibitors, preparation of antitumor agents, intermolecular Friedel-Crafts acylation, preparation of inhibitors of cell division in E. coli and synthesis of Hantzsch pyridine-containing Schiff bases.

    Solubility
    Soluble in ethanol and acetone. Insoluble in water.

    Notes
    Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit