Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indole-4-carboxaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Indolecarbaldehyde is a synthetic intermediate used for pharmaceutical synthesis. Also used as reactant in Biginelli reaction, synthesis of aurora kinase A inhibitors, preparation of antitumor agents, intermolecular Friedel-Crafts acylation, preparation of inhibitors of cell division in E. coli and synthesis of Hantzsch pyridine-containing Schiff bases.
Solubility
Soluble in ethanol and acetone. Insoluble in water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
4-Indolecarbaldehyde is a synthetic intermediate used for pharmaceutical synthesis. Also used as reactant in Biginelli reaction, synthesis of aurora kinase A inhibitors, preparation of antitumor agents, intermolecular Friedel-Crafts acylation, preparation of inhibitors of cell division in E. coli and synthesis of Hantzsch pyridine-containing Schiff bases.
Solubility
Soluble in ethanol and acetone. Insoluble in water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 143°C to 144°C |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in ethanol and acetone. Insoluble in water. |
Formula Weight | 145.16 |
Percent Purity | 97% |
Chemical Name or Material | Indole-4-carboxaldehyde |
Safety and Handling
Hazard Category | H302-H317-H319 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction