Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-5-carboxylic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1670-81-1
C9H7NO2
161.16
MFCD00005678
IENZCGNHSIMFJE-UHFFFAOYSA-N
indole-5-carboxylic acid,5-carboxyindole,5-indolecarboxylic acid,indole-5-carboxylic aicd,pubchem1694,5-carboxy-1h-indole,indole-5-carboxylicacid,indol-5-carboxylic acid,5-indole carboxylic acid,indole 5-carboxylic acid
74280
1H-indole-5-carboxylic acid
C1=CC2=C(C=CN2)C=C1C(=O)O
1670-81-1
C9H7NO2
161.16
MFCD00005678
IENZCGNHSIMFJE-UHFFFAOYSA-N
indole-5-carboxylic acid,5-carboxyindole,5-indolecarboxylic acid,indole-5-carboxylic aicd,pubchem1694,5-carboxy-1h-indole,indole-5-carboxylicacid,indol-5-carboxylic acid,5-indole carboxylic acid,indole 5-carboxylic acid
74280
1H-indole-5-carboxylic acid
C1=CC2=C(C=CN2)C=C1C(=O)O
Quantity 1g
Packaging Glass bottle
Color Beige to Yellow
Infrared Spectrum Authentic
Melting Point 207.0°C to 212.0°C
Assay Percent Range 98%
Formula Weight 161.16
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Indole-5-carboxylic acid, 98%

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-799-6