Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-6-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
1196-70-9
C9H7NO
145.16
MFCD02179596
VSPBWOAEHQDXRD-UHFFFAOYSA-N
indole-6-carboxaldehyde,6-formylindole,1h-indole-6-carboxaldehyde,indole-6-carbaldehyde,6-indolecarboxaldehyde,indole-6-carboxalehyde,6-formyl-1h-indole,indole-6-carboxaldehyde 6-formylindole,6-formyl-indole,6-formyl indole
2773435
1H-indole-6-carbaldehyde
C1=CC(=CC2=C1C=CN2)C=O
1196-70-9
C9H7NO
145.16
MFCD02179596
VSPBWOAEHQDXRD-UHFFFAOYSA-N
indole-6-carboxaldehyde,6-formylindole,1h-indole-6-carboxaldehyde,indole-6-carbaldehyde,6-indolecarboxaldehyde,indole-6-carboxalehyde,6-formyl-1h-indole,indole-6-carboxaldehyde 6-formylindole,6-formyl-indole,6-formyl indole
2773435
1H-indole-6-carbaldehyde
C1=CC(=CC2=C1C=CN2)C=O
Melting Point 129°C to 131°C
Infrared Spectrum Authentic
Color Brown to Yellow
Packaging Glass bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 145.16
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Indole-6-carboxaldehyde

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning