Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
120-72-9
C8H7N
117.151
MFCD00005607
SIKJAQJRHWYJAI-UHFFFAOYSA-N
indole,indol,2,3-benzopyrrole,1-benzazole,ketole,1-azaindene,benzopyrrole,2,3-benzopyrole,caswell no. 498b,indol german
798
CHEBI:16881
1H-indole
C1=CC=C2C(=C1)C=CN2
120-72-9
C8H7N
117.151
MFCD00005607
SIKJAQJRHWYJAI-UHFFFAOYSA-N
indole,indol,2,3-benzopyrrole,1-benzazole,ketole,1-azaindene,benzopyrrole,2,3-benzopyrole,caswell no. 498b,indol german
798
CHEBI:16881
1H-indole
C1=CC=C2C(=C1)C=CN2
Density 1.2200g/mL
Quantity 500g
Color Pink to White
Melting Point 51°C to 53°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 20, 304
Merck Index 15, 5003
Formula Weight 117.15
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material Indole, 98+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-420-7

RTECSNumber : NL2450000

TSCA : TSCA