Learn More
Alfa Aesar™ Indolicidin
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J65215EXD
 View more versions of this product
Catalog No. AAJ65215EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1906.28
0.5mg
Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2
-30° to -10°C
HPLC
Powder
RUO
Indolicidin