Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
496-15-1
C8H9N
119.17
MFCD00005705
LPAGFVYQRIESJQ-UHFFFAOYSA-N
indoline,2,3-dihydroindole,1-azaindan,dihydroindole,1h-indole, 2,3-dihydro,azaindane,benzopyrrolidine,aza-indane,indoline, 19,pubchem7502
10328
CHEBI:43295
2,3-dihydro-1H-indole
C1CNC2=CC=CC=C21
496-15-1
C8H9N
119.17
MFCD00005705
LPAGFVYQRIESJQ-UHFFFAOYSA-N
indoline,2,3-dihydroindole,1-azaindan,dihydroindole,1h-indole, 2,3-dihydro,azaindane,benzopyrrolidine,aza-indane,indoline, 19,pubchem7502
10328
CHEBI:43295
2,3-dihydro-1H-indole
C1CNC2=CC=CC=C21
Specific Gravity 1.063
Flash Point 92°C
Boiling Point 220°C to 221°C
Melting Point -21°C
Refractive Index 1.5920 to 1.594
Density 1.0630g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
Color Brown to Colorless
Assay Percent Range 97.5% min. (GC)
Beilstein 20, 257
Solubility Information Solubility in water: 5g/L (20°C)
Formula Weight 119.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Indoline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-816-8