Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indomethacin, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
53-86-1
C19H16ClNO4
357.79
MFCD00057095
CGIGDMFJXJATDK-UHFFFAOYSA-N
indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon
3715
CHEBI:49662
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
53-86-1
C19H16ClNO4
357.79
MFCD00057095
CGIGDMFJXJATDK-UHFFFAOYSA-N
indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon
3715
CHEBI:49662
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
Solubility
Soluble in acetone (40 mg/mL - clear, yellow solution), ethanol (20 mg/mL), ether, castor oil; Soluble in chloroform (50 mg/mL - clear, yellow, extremely viscous solution); decomposed by strong alkali but stable in neutral or slightly acidic media; insoluble in water.

Notes
Store away from oxidizing agents, strong bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Sensitivity Light sensitive
Melting Point 158°C to 161°C
Quantity 5g
Assay Percent Range 0.98
UN Number UN2811
Beilstein 497341
Merck Index 14,4968
Solubility Information Soluble in acetone (40mg/mL- clear,yellow solution),ethanol (20mg/mL),ether,castor oil; Soluble in chloroform (50mg/mL- clear,yellow,extremely viscous solution); decomposed by strong alkali but stable in neutral or slightly acidic media; insoluble in water.
Formula Weight 357.8
Percent Purity 98%
Chemical Name or Material Indomethacin
Show More Show Less

GHS H Statement
H300-H341

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P308+P313-P330-P333+P313-P363-P501c

H300-H317-H360FD

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 200-186-5

RTECSNumber : NL3500000

TSCA : Yes

Recommended Storage : Ambient temperatures