Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indomethacin, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
53-86-1
C19H16ClNO4
357.79
CGIGDMFJXJATDK-UHFFFAOYSA-N
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid
COC1=CC=C2N(C(=O)C3=CC=C(Cl)C=C3)C(C)=C(CC(O)=O)C2=C1
53-86-1
C19H16ClNO4
357.79
CGIGDMFJXJATDK-UHFFFAOYSA-N
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid
COC1=CC=C2N(C(=O)C3=CC=C(Cl)C=C3)C(C)=C(CC(O)=O)C2=C1

IN126, 53-86-1, C19H16NO4Cl

Spectrum™ Indomethacin, USP is used to relieve pain, swelling, and joint stiffness caused by arthritis, gout, bursitis, and tendonitis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Loss on Drying 0.005
Ignition Residue 0.002
Quantity 25 kg
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%