Learn More
Pitsco Education INDOOR BALLOON TESTER
Supplier: Pitsco Education W44474
Before the big launch outside, wouldn’t it be nice to test student balloons in the confines of your school? This innovative launcher will test-fly hot-air balloons safely and effectively. Because the unit runs on electricity, you don’t have to worry about fumes from the unit. You will, however, need to have a heavy extension cord available (12 gauge recommended) for optimum performance. This unit is intended for indoor use only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.