Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoxyl-β-D-glucuronide cyclohexylammonium salt, Acros Organics

Click to view available options
Quantity:
50mg
35804-66-1
C20H28N2O7
408.451
YGIPNZMTCTUCAX-CYRSAHDMSA-N
cyclohexylammonium ion 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-1h-indol-3-yloxy oxane-2-carboxylate,3-indolyl-beta-d-glucuronide cyclohexylammonium salt,3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
71307134
cyclohexylazanium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(1H-indol-3-yloxy)oxane-2-carboxylate
C1CCC(CC1)[NH3+].C1=CC=C2C(=C1)C(=CN2)OC3C(C(C(C(O3)C(=O)[O-])O)O)O
35804-66-1
C20H28N2O7
408.451
YGIPNZMTCTUCAX-CYRSAHDMSA-N
cyclohexylammonium ion 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-1h-indol-3-yloxy oxane-2-carboxylate,3-indolyl-beta-d-glucuronide cyclohexylammonium salt,3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
71307134
cyclohexylazanium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(1H-indol-3-yloxy)oxane-2-carboxylate
C1CCC(CC1)[NH3+].C1=CC=C2C(=C1)C(=CN2)OC3C(C(C(C(O3)C(=O)[O-])O)O)O
Quantity 50mg
Formula Weight 408.45
Chemical Name or Material Indoxyl-β-D-glucuronide cyclohexylammonium salt