Learn More
Infectious Bursal Disease Virus Mouse anti-Virus, Alexa Fluor™ 350, Clone: IBDV9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237745AF350
Catalog No. NP23774A30
Add to cart
Description
Description
Infectious Bursal Disease Virus Monoclonal antibody specifically detects Infectious Bursal Disease Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
Infectious Bursal Disease Virus
IBDV9
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
Purified, Conjugated
Mouse
IgG2a
0.1 mL
Primary
VP3 protein of IBD virus
Western Blot, ELISA, Immunohistochemistry
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
IBDV
Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified very virulent IBD virus (field strain) derived from infected chicken bursas.
Protein A purified
Store at 4°C in the dark.
Monoclonal
Virus