Learn More
Infectious Bursal Disease Virus Mouse anti-Virus, Alexa Fluor™ 594, Clone: IBDV9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237745AF594
Catalog No. NP23774A94
Add to cart
Description
Description
Infectious Bursal Disease Virus Monoclonal antibody specifically detects Infectious Bursal Disease Virus in Virus samples. It is validated for Western Blot,ELISA,Immunohistochemistry,ELISA.
Specifications
Specifications
Infectious Bursal Disease Virus
IBDV9
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
IBDV
Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified very virulent IBD virus (field strain) derived from infected chicken bursas.
0.1 mL
Primary
VP3 protein of IBD virus
Western Blot, ELISA, Immunohistochemistry
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Virus