Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Infectious Bursal Disease Virus Mouse anti-Virus, Alexa Fluor™ 750, Clone: IBDV9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP237745AF750

Catalog No. NP23774A70


Add to cart

Description

Description

Infectious Bursal Disease Virus Monoclonal antibody specifically detects Infectious Bursal Disease Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Infectious Bursal Disease Virus
IBDV9
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
Conjugated, Purified
Mouse
IgG2a
0.1 mL
Primary
VP3 protein of IBD virus
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
IBDV
Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified very virulent IBD virus (field strain) derived from infected chicken bursas.
Protein A purified
Store at 4°C in the dark.
Monoclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit