Learn More
ACCU-SCOPE™ INFINITY 8.3 MP CMOS Color Camera
SDP
Catalog No. 01189192 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
5 V
Catalog No. Voltage
01-189-192 5 V
1 options
Catalog No. 01-189-192 Supplier ACCU-SCOPE™ Supplier No. I8LC0008M
Add to Cart
Add to Cart
The new INFINITY 8-8C camera is designed to reproduce what your eye sees in the eyepiece, complete with an intuitive software package all for a price that will fit within the tightest budgets. Stream live videos or capture still images and share with ease.
  • Color CMOS sensor with 1/1.8 in. optical format
  • 8.3 megapixel camera with fast frame rates of 96 fps at full resolution
  • Direct USB3 interface for fast image delivery and simplified connectivity
  • Compact form factor
  • Support for locking industrial micro USB on the back for power and control
  • INFINITY ANALYZE microscopy software included with purchase. No annual license fees. (Camera requires INFINITY ANALYZE.7.0.3 or higher)
  • Software support for Windows and Mac, along with third-party plugins for MetaMorph and MicroManager
  • Selectable 8 or 12-bit pixel data
  • Recommended coupler: 0.5x to 0.63x
Specifications
Type USB Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 5 V