Learn More
Arlington Scientific Infinity Automated EIA Analyzer
Fully automated ELISA analyzer. Arlington Scientific Infinity Automated EIA Analyzer is a user-friendly, programmable open system which is capable of performing a single test or up to 12 various ELISA assays simultaneously in a full walkaway manner.
Manufacturer: Arlington Scientific 2700000
Description
- Programmable open system
- Walk-away automation
- Efficient control system manages operations, reporting and calibration functions
- Reproducible results
- Smart software automatically manages each assay separately; multiple assays can be processed on the same plate even if they have different timings, reagents and reading modes
Applications
EIA testing
Specifications
Automated EIA Analyzer | |
Performing a single test or up to 12 various ELISA assays simultaneously in a full walkaway manner |