Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza A H10N8 HA Monoclonal Antibody (mh001)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529946
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin/HA.
Specifications
| Influenza A H10N8 HA | |
| Monoclonal | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| Recombinant Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin/HA protein. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Inhibition Assays, Neutralization | |
| mh001 | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction