Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody, DyLight 755 - (A/USSR/90/1977), Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10062125IR
Description
Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza virusN1 samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Influenza A H1N1 Hemagglutinin | |
Polyclonal | |
50mM Sodium Borate | |
Influenza A, strain USSR (H1N1) | |
0.1 mL | |
Virology Bacteria and Parasites | |
Influenza virus N1 | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction