Learn More
Novus Biologicals™ Influenza A H5N1 Antibody Pair [Biotin]
Polyclonal/Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794295PLATES
 View more versions of this product
Catalog No. NB009898
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H5N1 (Avian Flu).
Specifications
Specifications
Biotin
Antibody Pairs
78.125 pg/mL
3654620
RUO
Influenza A H5N1
5 Plates
78.13-5000 pg/mL
1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to a working concentration of 1.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H5N1 (Avian Flu).
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL biotinylated rabbit anti-Influenza A H5N1 polyclonal antibody.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Influenza A H5N1 monoclonal antibody.
  • Standard: Each vial contains 150 ng of recombinant Influenza A H5N1.
  • Storage is content dependent.
    Rabbit/Mouse
    Viral
    HA, Hemagglutinin