Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H5N1 Mouse anti-Virus, Alexa Fluor™ 350, Clone: 2B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222605AF350

Catalog No. NP22260A30


Add to cart

Description

Description

Influenza A H5N1 Monoclonal antibody specifically detects Influenza A H5N1 in Virus samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Influenza A H5N1
2B7
Western Blot, ELISA
HA, Hemagglutinin
Recombinant human H5N1/HA1 (17-338aa) purified from Baculovirus
0.1 mL
Primary
3654620
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit