Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Influenza A Haemagglutinin H1N1 ELISA Kit (Colorimetric)

Recognizes both recombinant and natural Influenza A Haemagglutinin H1N1

$629.00

Specifications

Product Type ELISA Kit (Colorimetric)
Quantity 1 Kit
Sample Type Protein
Storage Requirements Storage is content dependent.
Target Species Influenza A Haemagglutinin H1N1
View More Specs

Products 2
Catalog Number Mfr. No. Assay Range Assay Sensitivity Price Quantity & Availability  
Catalog Number Mfr. No. Assay Range Assay Sensitivity Price Quantity & Availability  
NB013404
View Documents
Novus Biologicals™
NBP280333
31.25 to 2000 pg/mL 14.87 pg/mL
Each for $629.00
Add to cart
 
NB013421
View Documents
Novus Biologicals™
NBP280350
46.88 to 3000 pg/mL 10 pg/mL
Each for $629.00
Add to cart
 
Description

Description

Specifications

Specifications

ELISA Kit (Colorimetric)
Protein
Influenza A Haemagglutinin H1N1
1 Kit
Storage is content dependent.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit