Learn More
Influenza A Haemagglutinin H7 Mouse, Alexa Fluor 700, Clone: InA414, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150817AF700
Catalog No. NBP150817AI
Add to cart
Description
Description
Influenza A Haemagglutinin H7 Monoclonal antibody specifically detects Antigen in Human, Virus samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications
Specifications
Influenza A Haemagglutinin H7
InA414
Western Blot, ELISA
Purified
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A H7N1 or with Influenza A/Netherlands/219/03 H7N7 virus derived from allantoic fluid of 10 days old embryonated eggs
Protein G purified
RUO
Primary
Human, Virus
ELISA, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2b
0.25 ml
Store at 4C in the dark.
Monoclonal