Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A Hemagglutinin H7N7 Rabbit anti-Virus, Clone: 19, Novus Biologicals™
Rabbit Monoclonal Antibody
$493.00 - $666.00
Specifications
Antigen | Influenza A Hemagglutinin H7N7 |
---|---|
Clone | 19 |
Dilution | ELISA 0.5-1 ug/ml, Block/Neutralize |
Applications | ELISA, Neutralization |
Classification | Monoclonal |
Description
Influenza A Hemagglutinin H7N7 Monoclonal specifically detects Influenza A Hemagglutinin H7N7 in Virus samples. It is validated for ELISA, Block/Neutralize.Specifications
Influenza A Hemagglutinin H7N7 | |
ELISA 0.5-1 ug/ml, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
IgG | |
Protein A purified |
19 | |
ELISA, Neutralization | |
Unconjugated | |
Rabbit | |
Virus | |
Recombinant Influenza A Hemagglutinin H7N7 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title