Learn More
Alfa Aesar™ Influenza A NP (366-374)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66443MB
 View more versions of this product
Catalog No. AAJ66443MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
982.05
25mg
Ala-Ser-Asn-Glu-Met-Asn-Asp-Ala-Met
-30° to -10°C
HPLC
Solid
RUO
Influenza A NP (366-374)