Learn More
Influenza B virus Goat anti-Influenza virus, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29792
Catalog No. 89-033-637
Add to cart
Description
Description
Goat polyclonal antibody raised against influenza B virus.
  • High specificity
  • High reactivity
Specifications
Specifications
Influenza B virus
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Influenza B virus (Yamagata strain).
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Influenza virus
ELISA, Immunofluorescence, Western Blot
Goat polyclonal antibody raised against influenza B virus.
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Goat
IgG
RUO
Primary
This antibody reacts with influenza B virus (Yamagata strain) virions. Does not cross-react with influenza A virus, RSV, parainfluenza virus type 1,2,3 or adenovirus.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)