Learn More
Influenza B virus Mouse anti-Influenza virus, Clone: J335, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13604
Catalog No. 89-033-567
Add to cart
Description
Description
Mouse monoclonal antibody raised against influenza B virus.
  • High specificity
  • High reactivity
Specifications
Specifications
Influenza B virus
J335
Mouse monoclonal antibody raised against influenza B virus.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with Nucleoprotein of influenza B virus.
Immunofluorescence, Lateral Flow
Unconjugated
Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2b
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Influenza virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)