Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B Virus Nucleoprotein Mouse anti-Virus, Alexa Fluor™ 750, Clone: 1D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223514AF750

Catalog No. NP22314A70


Add to cart

Description

Description

Influenza B Virus Nucleoprotein Monoclonal antibody specifically detects Influenza B Virus Nucleoprotein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Western Blot.
Specifications

Specifications

Influenza B Virus Nucleoprotein
1D8
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Western Blot (Negative)
Conjugated, Purified
Mouse
IgG2a Kappa
0.1 mL
Primary
Clone 1D8 reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Clone 1D8 does not cross-react with influenza A virus.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Influenza B Virus NP, NP, Nucleoprotein
Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37C.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit