Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Virus Nucleoprotein Mouse anti-Virus, Clone: 1B6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105187
Description
Influenza B Virus Nucleoprotein Monoclonal antibody specifically detects Influenza B Virus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Influenza B Virus Nucleoprotein | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:40, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000 | |
Mouse | |
Protein A or G purified | |
RUO | |
Reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Does not cross-react with influenza A virus (2) | |
Store at 4C. Do not freeze. | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1B6 | |
Unconjugated | |
Influenza B Virus NP, NP, Nucleoprotein | |
Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37 degrees C. | |
100 μg | |
Primary | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only