Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Virus Nucleoprotein Mouse anti-Virus, Clone: 1B6, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Influenza B Virus Nucleoprotein |
---|---|
Clone | 1B6 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:40, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105187
|
Novus Biologicals
NBP105187 |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018350
|
Novus Biologicals
NBP1051870.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Influenza B Virus Nucleoprotein Monoclonal antibody specifically detects Influenza B Virus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Influenza B Virus Nucleoprotein | |
1.0 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Virus | |
Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37 degrees C. | |
Primary | |
Store at 4C. Do not freeze. |
1B6 | |
Western Blot 1:40, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Influenza B Virus NP, NP, Nucleoprotein | |
IgG2a κ | |
Protein A or G purified | |
Reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Does not cross-react with influenza A virus (2) |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title