Learn More
InfluenzaAH3N2 Goat anti-Influenza virus, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP266599
Catalog No. NBP266599
Add to cart
Description
Description
InfluenzaAH3N2 Polyclonal antibody specifically detects InfluenzaAH3N2 in Influenza virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
InfluenzaAH3N2
Unconjugated
PBS (pH 7.5) before the addition of 40% glycerol with 0.1% Sodium Azide
Goat
IgG
1mg
Primary
Influenza virus
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
Influenza A, H3N2 strain
>95%
Store at 4°C after rehydration. For extended storage after rehydration, add an equal volume of glycerol and store at -20°C. Avoid repeated freeze-thaw cycles.
Polyclonal