Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InfluenzaAHaemagglutininH1N1 Mouse anti-Influenza A Virus H1, Clone: 8L383, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266619

Catalog No. NBP266619


Add to cart

Description

Description

InfluenzaAHaemagglutininH1N1 Monoclonal antibody specifically detects InfluenzaAHaemagglutininH1N1 in Influenza A Virus H1 samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

InfluenzaAHaemagglutininH1N1
8L383
ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Purified
Mouse
IgG1
100μg
Primary
Influenza A Virus H1
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
PBS with 0.1% Sodium Azide
HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin
BALB/c mice immunized with purified influenza virus type A strain H1N1.
Protein A or G purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit