Learn More
InfluenzaBVirusNucleoprotein Mouse anti-Virus, Janelia Fluor 646, Clone: 1B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105187JF646
Catalog No. NBP15187646
Add to cart
Description
Description
InfluenzaBVirusNucleoprotein Monoclonal antibody specifically detects InfluenzaBVirusNucleoprotein in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
InfluenzaBVirusNucleoprotein
1B6
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG2a Kappa
0.1mL
Primary
Reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Does not cross-react with influenza A virus (2)
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Influenza B Virus NP, NP, Nucleoprotein
Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37 degrees C.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus